Anti-HNF-1beta Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB137137-100
BIRBORB137137-100EA 0 GBP
BIRBORB137137-100
Anti-HNF-1beta Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:1000

Type: Primary
Antigen: HNF-1beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR