Anti-DNMT3B Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB135397-100 ORB135397-200 ORB135397-50
BIRBORB135397-100EA 0 GBP
BIRBORB135397-100 BIRBORB135397-200 BIRBORB135397-50
Anti-DNMT3B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNMT3B (DNA (cytosine-5-)-methyltransferase 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol DNMT3B DNMT3B
Environmentally Preferable
Antigen name DNA (cytosine-5-)-methyltransferase 3 beta DNA (cytosine-5-)-methyltransferase 3 beta
Antibody type Primary Primary
Clonality Polyclonal Polyclonal
Conjugation Unconjugated Unconjugated
Reactivity Human,Rat,Mouse Human,Rat,Mouse
Host Rabbit Rabbit
Isotype IgG IgG
Western blot Yes Yes
Purification purified by affinity chromatography
Storage temperature Cool Cool
Shipping temperature Ice Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR