Anti-ALOX12 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB124918-100
BIRBORB124918-100EA 0 GBP
BIRBORB124918-100
Anti-ALOX12 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)
Antibodies
Type: Primary
Antigen: ALOX12 (arachidonate 12-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol ALOX12
Antigen name Arachidonate 12-lipoxygenase
Environmentally Preferable
Antigen synonyms 12-LOX
Antibody type Primary
Clonality Polyclonal
Conjugation PE (Phycoerythrin)/Cy3®
Reactivity
Host Rabbit
Isotype IgG
Immunogen KLH conjugated synthetic peptide derived from human 12 Lipoxygenase.
Form liquid
Molecular weight 73 kDa
Purification purified by affinity chromatography
Storage buffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR