Intestinal villi model
Supplier: 3B Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1005532
BINH1005532EA 0 GBP
BINH1005532
Intestinal villi model
Models Human Biology Models
This intestinal villi model consists of one entire villus, one longitudinally sectioned villus showing the arterioles and venules and one sectioned villus to show the lymphatic vessels. The intestinal villi model also includes a longitudinal section of Lieberkühn's crypt.
  • Magnified 100 times
  • On base