Chairs and stools, Koncept
Supplier: Kango
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7NK90NBDU00905 7NK90NHDP01905 7NK91NBDU00905 7NK91NHDP01905 7NK93VHXX00905 7NK95VHXX00905 7NK95VHXX04905 7NK90NBDV00905 7NK90NBDR00905 7NK90NBDP00905 7NK90NBDC00905 7NK91NBDR00905 7NK91NBDC00905 7NK97GPDH00905 7NK91NHDP00905 7NK91NCDR00905 7NK91NBDV00905 7NK91NBDP00905 7NK97GPDU00905 7NK90NHDP00905 5NK90NBDP00P95 5NK90NBDC00P95 5NK90NBDR00P95 5NK90NBDU00P95 5NK90NBDV00P95 5NK90NHDP00P95 5NK90NHDP01P95 5NK91NBDP00P95 5NK91NBDC00P95 5NK91NBDR00P95 5NK91NBDU00P95 5NK91NBDV00P95 5NK91NHDP00P95 5NK91NHDP01P95 7NK93VHXX04905
139-0447EA 344 GBP
139-0447 139-0442 139-1017 139-1018 139-0457 BIEN7NK95VHXX00905 139-1019 139-0448 139-0441 139-0446 139-0445 139-0443 139-0453 BIEN7NK97GPDH00905 139-0456 BIEN7NK91NCDR00905 139-0455 139-0454 139-0458 139-0449 BIEN5NK90NBDP00P95 BIEN5NK90NBDC00P95 BIEN5NK90NBDR00P95 BIEN5NK90NBDU00P95 BIEN5NK90NBDV00P95 BIEN5NK90NHDP00P95 BIEN5NK90NHDP01P95 BIEN5NK91NBDP00P95 BIEN5NK91NBDC00P95 BIEN5NK91NBDR00P95 BIEN5NK91NBDU00P95 BIEN5NK91NBDV00P95 BIEN5NK91NHDP00P95 BIEN5NK91NHDP01P95 BIEN7NK93VHXX04905
Chairs and stools, Koncept
Chairs
Ergonomic and auto-adaptive chairs or stools made from soft PU or wood.
Nobody has perfect symmetrical morphology. This is why people have back problems, sometimes with a feeling of heavy legs. The answer to this problem can be found in the main features of the Koncept chairs and stools.
  • Auto-adaptive to users and to their positions
  • Helps the mobility, blood flow and elimintates back pain
  • No staples or seams
  • Abrasion-resistant
  • Black steel five-branch scuff resistant reinforced base
  • Black gas lift column with height adjustment and shock absorber (except for
More Product Information