Burettes, removable stopcock
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BRB276P BRB284 BRB290 BRB264P BRB266P
BIBBBRB276PEA 25 GBP
BIBBBRB276P BIBBBRB284 612-0376 612-0369 612-0370
Burettes, removable stopcock
Burettes
Soda lime glass, class B, removable PTFE stopcock.
  • Flared top for safer filling
  • Blue enamel graduations
BS 846
Certifications: BS 846