Homogeniser, Dounce Type
Supplier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1984-10007 1984-10100 1984-10040 1984-10002
BELC1984-10007EA 226.18 GBP
BELC1984-10007 BELC1984-10100 BELC1984-10040 BELC1984-10002
Homogeniser, Dounce Type
Homogenisers Tissue Grinders
Dounce tissue homogeniser designed for disruption of whole cells leaving cell nuclei undamage.
  • Polished tube and pestles
  • Made up of glass
Delivery information: Two pestles supplied with each unit, large clearance pestle A for initial processing and small clearance pestle B completes homogenisation.