Pipette canisters, round bottom
Supplier: BELLCO GLASS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1287-00430 1287-38250 1287-38430
BELC1287-00430EA 153.38 GBP
BELC1287-00430 BELC1287-38250 BELC1287-38430
Pipette canisters, round bottom
Jars Pipette Jars
Pipette canister made of borosilicate glass, class A, type I.
  • Manufactured from Type 1, Class A borosilicate glass
  • Components will withstand repeated dry or steam heat sterilisation