Separating funnel stands, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H189640000
BELAH189640000EA 602 GBP
BELAH189640000
Separating funnel stands, SP Bel-Art
Retort Stands
This rotary separatory funnel rack consists of a rectangular base, screw-in column and a support disc held in place by a screw clamp.
  • Support disc is fully adjustable for different heights up to 21"
  • Open front type support holes for easy removal and placement of separatory funnels
  • Shipped unassembled
Ordering information: Includes 18 insert clips to accommodate different sized funnels.