Drawer organiser trays, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H186550000 H186540000 H186520000
BELAH186550000EA 273 GBP
BELAH186550000 BELAH186540000 BELAH186520000
Drawer organiser trays, SP Bel-Art
Trays/Dishes/Pots Drawer Organisation Trays
Drawer organiser, made of white polystyrene.
  • Fits into standard laboratory bench drawers
  • Easy to clean
Various configurations available to accommodate thermometers, medication cups, jars, stir bars, vials, and pipettes.