Vacuum desiccators, Lab Companion™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F424002021 F424002221 F424002141 F424002121 F424002041
BELAF424002021EA 844 GBP
BELAF424002021 BELAF424002221 BELAF424002141 BELAF424002121 BELAF424002041
Vacuum desiccators, Lab Companion™, SP Bel-Art
Desiccators Desiccators, Plastic
These compact, high-quality vacuum desiccators in clear or amber (UV blocking) polycarbonate maintain a vacuum and stay gas-tight allowing reliable dry storage or experiments over extended periods.
  • Able to maintain 29 inches Hg vacuum for over 72 hours at room temperature
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage
  • Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release or gas exchange
  • Easy to clean, chemical resistant and allow quick visualisation of
More Product Information