Desiccator cabinets, vertical, secador®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F420741000 F420721000 F420710000 F420731008 F420741008 F420710008 F420731000
BELAF420741000EA 2340 GBP
BELAF420741000 BELAF420721000 BELAF420710000 BELAF420731008 BELAF420741008 BELAF420710008 BELAF420731000
Desiccator cabinets, vertical, secador®, SP Bel-Art
Desiccators Desiccator Cabinets
Dessicator cabinets which can be used with disposable or reusable Scienceware® desiccant for safe storage of moisture sensitive products. Desiccant can be placed on or under the perforated bottom shelf. The door opening is 11,4×22,5 cm and includes latch and 2 loops for seals.
  • Portable cabinets have convenient built in carrying handles
  • Vertical cabinets are interchangably stackable
  • Door can be secured with tamper evident tags or padlock
  • Easy to view dial hygrometer in the door