Magnetic stirring bar retrievers, PTFE, jumbo, Spinbar®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F377810000
BELAF377810000EA 630 GBP
BELAF377810000
Magnetic stirring bar retrievers, PTFE, jumbo, Spinbar®, SP Bel-Art
Stirrers Stirring Tools
Flexible PTFE magnetic retriever for removing the largest magnetic stirring bars from deep vessels. Designed with powerful Alnico 5 magnetic end having 102 mm in length.
  • Easy grip 'T' handle
  • Longer, custom lengths are available