Glove box holders, Poxygrid®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F247390002 F247390003 F247390001
BELAF247390002EA 158 GBP
BELAF247390002 BELAF247390003 BELAF247390001
Glove box holders, Poxygrid®, SP Bel-Art
Glove Dispensers and Glove Box Holders
Epoxy-coated, welded steel wire racks for one, two or three boxes of disposable gloves, wipes or tissues.
  • Glove box drops into an individual retaining compartment and lifts out easily for replacement
  • Designed for mounting to a vertical surface
  • Durable, blue epoxy coated racks are steam autoclavable at 121 °C