Dryers for laboratory glassware, benchtop or wallmount, Lab-Aire® II, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F189330011 F189330012 F189330014 F189330015 F189330022 F189330013
BELAF189330011EA 270 GBP
BELAF189330011 BELAF189330012 BELAF189330014 BELAF189330015 BELAF189330022 BELAF189330013
Dryers for laboratory glassware, benchtop or wallmount, Lab-Aire® II, SP Bel-Art
Racks Draining Racks
Non-electric benchtop models let the user places the dryer wherever it will be most convenient.
  • Sturdy base provides excellent balance and support
  • Single-sided models fit nicely against a wall for economy of space
  • Turn double-sided models perpendicular to the bench for easy access to both sides
  • Wallmount dryers keep labware up off the bench and out of the way as it dries
  • Easily installs on walls or standard pegboard using molded-in tabs with key hole openings