Digestion system, behrotest®
Supplier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B00218448 B00218430 B00218446
BEHRB00218448EA 3300 GBP
BEHRB00218448 BEHRB00218430 BEHRB00218446
Digestion system, behrotest®
Digestion Systems Mineralisation Workstations
Digestion apparatus for crude fibre or hydrolysis.
Delivery information: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
Order Now
SPECIFICATIONS
Environmentally Preferable