Sampling scoops, Behrotest®
Supplier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B00196081 B00196079 B00196080
BEHRB00196081EA 590 GBP
BEHRB00196081 331-3110 331-3120
Sampling scoops, Behrotest®
Samplers Water Samplers
The sampling scoop is composed of a clamping fixture and end bar. The scoop with receptacles made of borosilicate glass comply with the high requirements.
  • Sturdy three-part supporting bar with telescopic racks
  • Continuously adjustable in length
Suitable for sampling in waters that are difficult to access, from the surface.
Certifications: EN 1485/AOX