CytoFLEX platform flow cytometers
Supplier: Beckman Coulter
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B53010 B53011 B53012 B53016 B53000 B53001 B53002 B53003 B53004 B53005 B53006 B53007 B53008 B53009 B53013 B53014 B53018 B53037 B96622 C02944 B75408 B75811 B75812 B78557 B78558 B78559 B78560 C02945 C02946 B78561 B96618 B96619 B96620 B96621 C02948 C00445 C00446 C02949 C11183 C11184 C01158 C01159 C01160 C01161 C02947 C11185 C11186
BECMB53010EA 0 GBP
BECMB53010 BECMB53011 BECMB53012 BECMB53016 BECMB53000 BECMB53001 BECMB53002 BECMB53003 BECMB53004 BECMB53005 BECMB53006 BECMB53007 BECMB53008 BECMB53009 BECMB53013 BECMB53014 BECMB53018 BECMB53037 BECMB96622 BECMC02944 BECMB75408 BECMB75811 BECMB75812 BECMB78557 BECMB78558 BECMB78559 BECMB78560 BECMC02945 BECMC02946 BECMB78561 BECMB96618 BECMB96619 BECMB96620 BECMB96621 BECMC02948 BECMC00445 BECMC00446 BECMC02949 BECMC11183 BECMC11184 BECMC01158 BECMC01159 BECMC01160 BECMC01161 BECMC02947 BECMC11185 BECMC11186
CytoFLEX platform flow cytometers
Cell Counters and Flow Cytometers Flow Cytometers
The CytoFLEX platform flow cytometer is a revolutionary system presenting optimal excitation and emission, minimising light loss and maximising sensitivity.
  • Exquisite sensitivity (80 nm) enabling nano particle analysis including extracellular vesicles, viruses and more
  • Extensive set of repositionable band pass filters
  • Flexibility to upgrade by adding additional parameters
  • Intuitive software to facilitate multicolour analysis
The CytoFLEX platform is divided into several versions across three models. An instrument is defined by the lasers included.More Product Information