Kaluza Analysis Software Perpetual, 1-User (Single)
Supplier: Beckman Coulter
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A82959
BECMA82959EA 4590 GBP
BECMA82959
Kaluza Analysis Software Perpetual, 1-User (Single)
Software Laboratory Software
Kaluza analysis can load any listmode file that is compliant with the FCS standard up through version 3.1.
  • License type: Individual user
  • Number of users: 1
  • License term: Perpetuity
Kaluza Analysis Software is a stand-alone program supporting data analysis from any Flow Cytometry Standard (FCS)-compliant file, regardless of the manufacturer. This is a full-featured high-performance package with improved processing capabilities for multidimensional analysis including enhanced color compensation algorithms and real time adjustments to data displayMore Product Information