BD Venflon™ Catheters IV
Supplier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
391452 391451 391455 391453 391457 391454 381844 391456
BDAM391452EA 229 GBP
BDAM391452 BDAM391451 BDAM391455 BDAM391453 BDAM391457 BDAM391454 BDAM381844 BDAM391456
BD Venflon™ Catheters IV
Catheters
Intravenous catheters with injection port and valve for peripheral catheterisation allows easier insertion and minimises the risk of vein damage.
  • Permits needle-free administration of bolus medications during infusion
  • Back-cut needle bevel and tapered end of the PTFE catheter
  • Wing position reduces the risk of bacterial contamination
  • Prevent contact between the catheter hub and the skin
  • Using the wings, the catheter can be maintained with the BD Veca-C™ and BD Veca-Fix™ IV dressing
BD Luer-Lok™ compatible.