Angiocath™ IV Catheters
Supplier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
382277 382259 381164 382258 382268 381112 381123 381134 381137 381144 381147 381154 381157 382287
BDAM382277EA 161 GBP
BDAM382277 BDAM382259 BDAM381164 BDAM382258 BDAM382268 BDAM381112 BDAM381123 BDAM381134 BDAM381137 BDAM381144 BDAM381147 BDAM381154 BDAM381157 BDAM382287
Angiocath™ IV Catheters
Cannulae
The BD Angiocath™ and BD Angiocath™ special IV catheters offer a wide range of products designed to provide peripheral vascular access to help meet clinicians' needs. The catheters are made with FEP material with a clear flash chamber to help make blood flashback easily identifiable.
  • Made with a radiopaque FEP material that provides x-ray detectability
  • BD Angiocath™ special IV catheters help offer a wide range of gauges and lengths from 10 to 16 G in multiple length sizes