BD Vacutainer® UltraTouch™ Push Button Blood Collection Sets
Supplier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
367364 367365
BDAM367364EA 270 GBP
BDAM367364 BDAM367365
BD Vacutainer® UltraTouch™ Push Button Blood Collection Sets
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
BD Vacutainer® UltraTouch™ Push button blood collection set is engineered to minimize patient discomfort during blood collection.
  • Single-handed, in-vein safety activation instantly retracts needle after use
  • PentaPoint™ bevel requires 32% less penetration force
Ultra-thin RightGauge™ cannula allows the use of a smaller gauge suitable for more veins, without compromising filling times or sample quality.
Order Now
SPECIFICATIONS
Environmentally Preferable