Serum separator tubes, SST™ II advance, BD Vacutainer®
Supplier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
367957 367955 367953 367954 368879 366882 368969 368965 368966 368970
BDAM367957EA 74.5 GBP
BDAM367957 BDAM367955 BDAM367953 BDAM367954 BDAM368879 BDAM366882 BDAM368969 BDAM368965 BDAM368966 BDAM368970
Serum separator tubes, SST™ II advance, BD Vacutainer®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET, clear.
  • Silica (Clot Activator)/Gel
  • Stable barrier between serum and clotted blood, therefore better analyte stability
  • Better sample quality
  • Optimisation of the work flow: Short centrifugation time, sample processing and archiving in the primary tube
  • No possibility of misidentification due to the use of secondary tubes
During the centrifugation of the BD Vacutainer® SST™ II Advance tubes, an inert gel separates the serum and the blood clot preventing the contamination of the serum from the separated cellular components. For example, the serum forMore Product Information