Blood collection tubes, with ACD, BD Vacutainer®
Supplier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
367756 366645 366645
613-3989EA 139 GBP
613-3989 BDAM366645 BDAM366645
Blood collection tubes, with ACD, BD Vacutainer®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These ACD tubes are used to obtain a whole blood or a plasma sample.
  • Contains the active anticoagulant tri-sodium citrate with citric acid and dextrose
  • Available in two formulations, ACD solution A and ACD solution B
Tubes are available in glass only and are identifiable by a yellow cap.
Order Now
SPECIFICATIONS
Environmentally Preferable