Glycylglycine (Gly-Gly)

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4030664.0025 4030664.0100
BACE4030664.0025EA 0 GBP
BACE4030664.0025 BACE4030664.0100
Glycylglycine (Gly-Gly)
Proteins and Peptides

  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR