N-Decanoyl-N-methyl-D-glucamine (MEGA-10)

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4011133.0001 4011133.0005 4011133.0025
BACE4011133.0001EA 0 GBP
BACE4011133.0001 BACE4011133.0005 BACE4011133.0025
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
Formula: C₁₇H₃₅NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00036801
CAS Number: 85261-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR