N-Acetyl-D-methionine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4007232.0005
BACE4007232.0005EA 0 GBP
BACE4007232.0005
N-Acetyl-D-methionine
N-Acetyl-D-methionine
Formula: C₇H₁₃NO₃S
MDL Number: MFCD00069962
CAS Number: 1509-92-8
EINECS: 216-144-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR