L(+)-Ascorbic acid ≥99.5%, powder, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B581-05EA 84.6 GBP
B581-05
L(+)-Ascorbic acid ≥99.5%, powder, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
L(+)-Ascorbic acid

Powder. Lot analysis on label.

Formula: C₆H₈O₆
Boiling Pt: 553 °C (1013 hPa)
Melting Pt: 190…192 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 99.5 %
Assay (C₆H₈O₆) ≥ 99.5 %
Specific Rotation,α²⁵D(c=10 in water)(+) 20.5 - 21.5 Degree
Residue after Ignition ≤ 0.10 %
Loss on Drying at 105°C ≤ 0.1 %
Heavy Metals (as Pb) ≤ 0.001 %
Trace Impurities - Iron (Fe) ≤ 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR