Cyclobutylamine hydrochloride 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Cyclobutanamine hydrochloride

B25400.14 B25400.06
B25400.14EA 216 GBP
B25400.14 B25400.06
Cyclobutylamine hydrochloride 97%
Cyclobutylamine hydrochloride

Purity: 97% CAS No.: 6291-01-6 Molecular Formula: C4H9N.HCl Formula Weight: 107.58 Melting Point: 180-183° MDL No.: MFCD00034953

Formula: C₄H₉N·HCl
Melting Pt: 180…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00034953
CAS Number: 6291-01-6
EINECS: 228-540-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR