3-Amino-o-toluic acid 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B24947.06 B24947.14
B24947.06EA 18.4 GBP
B24947.06 B24947.14
3-Amino-o-toluic acid 98%
3-Amino-o-toluic acid

Purity: 98% CAS No.: 52130-17-3 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: 185-188° Beilstein Reference: 2803090

Formula: C₈H₉NO₂
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00075026
CAS Number: 52130-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR