Ethyl thiophene-2-glyoxylate ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B24712.18
B24712.18EA 84.6 GBP
B24712.18
Ethyl thiophene-2-glyoxylate ≥97%
Ethyl thiophene-2-glyoxylate

Purity: 97% CAS No.: 4075-58-5 Molecular Formula: C8H8O3S Formula Weight: 184.21 Flash Point: 110°(230°F) Refractive Index: 1.5505 Boiling Point: 102-110°/0.8mm Beilstein Reference: 5651 MDL No.: MFCD00015538

Formula: C₈H₈O₃S
Boiling Pt: 102…110 °C (0.8 mmHg)
Density: 1.250
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015538
CAS Number: 4075-58-5
EINECS: 223-793-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR