Aminoguanidinium hydrogen carbonate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B24696.0E B24696.22 B24696.36
B24696.0EEA 124 GBP
B24696.0E B24696.22 B24696.36
Aminoguanidinium hydrogen carbonate ≥98%
Aminoguanidinium hydrogen carbonate

Purity: 98+% CAS No.: 2582-30-1 Molecular Formula: CH6N4.H2CO3 Formula Weight: 136.11 Melting Point: ca 170° dec. Beilstein Reference: 3569869 UN No.: 3077 MDL No.: MFCD00012949

Formula: C₂H₈N₄O₃
Melting Pt: ∼ 170 °C (dec.)
Density: 1.600
Storage Temperature: Ambient
MDL Number: MFCD00012949
CAS Number: 2582-30-1
EINECS: 219-956-7
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR