4,4'-Dichlorobutyrophenone ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-Chloro-1-(4-chlorophenyl)butan-1-one, γ,4-Dichlorobutyrophenone, ω,4-Dichlorobutyrophenone

B24608.14 B24608.18 B24608.22
B24608.14EA 39 GBP
B24608.14 B24608.18 B24608.22
4,4'-Dichlorobutyrophenone ≥97%
4,4'-Dichlorobutyrophenone

Purity: 97% CAS No.: 40877-09-6 Molecular Formula: C10H10Cl2O Formula Weight: 217.09 Refractive Index: 1.5600 Melting Point: 28-32° Beilstein Reference: 879782 MDL No.: MFCD00018993

Formula: C₁₀H₁₀Cl₂O
Boiling Pt: 135…140 °C (0.9 mmHg)
Melting Pt: 28…32 °C
Density: 1.2575 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00018993
CAS Number: 40877-09-6
EINECS: 255-123-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR