2-Aminopyrimidine 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B24594.22 B24594.36
B24594.22EA 34 GBP
B24594.22 B24594.36
2-Aminopyrimidine 98%
2-Aminopyrimidine

Purity: 98% CAS No.: 109-12-6 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 123-127° Boiling Point: 159°/186mm MDL No.: MFCD00006089

Formula: C₄H₅N₃
Boiling Pt: 159 °C (186 mmHg)
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00006089
CAS Number: 109-12-6
EINECS: 203-648-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR