2,4-Dichloro-5-sulphamoylbenzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

B24589.18
B24589.18EA 37.6 GBP
B24589.18
2,4-Dichloro-5-sulphamoylbenzoic acid ≥98%
2,4-Dichloro-5-sulphamoylbenzoic acid

Purity: 98% CAS No.: 2736-23-4 Molecular Formula: C7H5Cl2NO4S Formula Weight: 270.09 Melting Point: 230-232° dec. MDL No.: MFCD00007931

Formula: C₇H₅Cl₂NO₄S
Melting Pt: 230…232 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007931
CAS Number: 2736-23-4
EINECS: 220-358-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR