Isoeugenol cis- and trans- mixture 98

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Propenylguaiacol, 4-Hydroxy-3-methoxy-1-propenylbenzene, 2-Methoxy-4-(prop-1-en-1-yl)phenol

B24541.14 B24541.22 B24541.36
B24541.14EA 19.8 GBP
B24541.14 B24541.22 B24541.36
Isoeugenol cis- and trans- mixture 98
Isoeugenol

Purity: 98+% CAS No.: 97-54-1 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 134°(273°F) Refractive Index: 1.5770 Melting Point: -10° Boiling Point: 266-268° Beilstein Reference: 1909602 Merck Reference: 14,5171 MDL No.: MFCD00009285

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 266…268 °C
Melting Pt: –10 °C
Density: 1.080
Flash Pt: 134 °C (273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009285
CAS Number: 97-54-1
EINECS: 202-590-7
Merck Index: 14,5171

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR