Methyl vanillate ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B24209.06 B24209.14 B24209.22
B24209.06EA 21.4 GBP
B24209.06 B24209.14 B24209.22
Methyl vanillate ≥99%
Methyl vanillate

Purity: 99% CAS No.: 3943-74-6 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 62-66° Boiling Point: 285-287° MDL No.: MFCD00008438

Formula: C₉H₁₀O₄
Boiling Pt: 285…287 °C
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00008438
CAS Number: 3943-74-6
EINECS: 223-525-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR