N-Methylisatoic anhydride tech. 90%, Technical Grade

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

B24099.22
B24099.22EA 56.3 GBP
B24099.22
N-Methylisatoic anhydride tech. 90%, Technical Grade
N-Methylisatoic anhydride

Purity: 90% CAS No.: 10328-92-4 Molecular Formula: C9H7NO3 Formula Weight: 177.16 Melting Point: ca 160° dec. MDL No.: MFCD00006815

Formula: C₉H₇NO₃
Melting Pt: ∼ 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006815
CAS Number: 10328-92-4
EINECS: 233-714-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR