(3-Chlorophenyl)methanesulphonyl chloride 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 3-Chlorobenzylsulphonyl chloride, 3-Chloro-α-toluenesulphonyl chloride

B24073.06 B24073.03 B24073.14
B24073.06EA 53.5 GBP
B24073.06 B24073.03 B24073.14
(3-Chlorophenyl)methanesulphonyl chloride 96%
(3-Chlorophenyl)methanesulphonyl chloride

Purity: 96% CAS No.: 24974-73-0 Molecular Formula: C7H6Cl2O2S Formula Weight: 225.09 Melting Point: 69-75° UN No.: 3261 MDL No.: MFCD02683111

Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
Melting Pt: 69…75 °C
Storage Temperature: Ambient
MDL Number: MFCD02683111
CAS Number: 24974-73-0
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR