2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol

B24048.14 B24048.03 B24048.06
B24048.14EA 312 GBP
B24048.14 B24048.03 B24048.06
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 96%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol

Purity: 96% CAS No.: 722-92-9 Molecular Formula: C9H7F6NO Formula Weight: 259.15 Melting Point: 147-152° Beilstein Reference: 1473737 MDL No.: MFCD00039258

Formula: C₉H₇F₆NO
MW: 259.151 g/mol
Melting Pt: 147…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039258
CAS Number: 722-92-9
EINECS: 670-530-0
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR