Butyl vinyl ether ≥98% stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: n-Butyl vinyl ether

Stabilisation: Stabilised with potassium hydroxide 100 ppm

B23673.AE B23673.AP
B23673.AEEA 23.3 GBP
B23673.AE B23673.AP
Butyl vinyl ether ≥98% stabilised
Butyl vinyl ether

Stabilized with 0.01% KOH


Purity: 98% CAS No.: 111-34-2 Molecular Formula: C6H12O Formula Weight: 100.16 Storage Temperature: 2°C to 8°C Flash Point: -12°(10°F) Refractive Index: 1.4010 Melting Point: -113° Boiling Point: 92-94° Beilstein Reference: 1560217 UN No.: 3271 MDL No.: MFCD00009454

Formula: C₆H₁₂O
Boiling Pt: 92…94 °C
Melting Pt: –113 °C
Density: 0.778
Flash Pt: –12 °C (10 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009454
CAS Number: 111-34-2
EINECS: 203-860-7
UN: 3271
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR