D-Norvaline ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B23444.03 B23444.06 B23444.14
B23444.03EA 17.7 GBP
B23444.03 B23444.06 B23444.14
D-Norvaline ≥99%
D-Norvaline

Purity: 99% CAS No.: 2013-12-9 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Melting Point: 300° Beilstein Reference: 1721161 Merck Reference: 14,6716 Optical Rotation: -25° (c=10 in 20% HCl) MDL No.: MFCD00008097

Formula: C₅H₁₁NO₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008097
CAS Number: 2013-12-9
EINECS: 217-936-2
Merck Index: 14,6716

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR