(1R,3S)-(+)-Camphoric acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B

B23106.14 B23106.22 B23106.36
B23106.14EA 22.8 GBP
B23106.14 B23106.22 B23106.36
(1R,3S)-(+)-Camphoric acid ≥98%
(1R,3S)-(+)-Camphoric acid

Purity: 98+% CAS No.: 124-83-4 Molecular Formula: C10H16O4 Formula Weight: 200.23 Melting Point: 183-188° Beilstein Reference: 2050204 Merck Reference: 14,1733 Optical Rotation: +46° (c=1 in methanol) MDL No.: MFCD00001375

Formula: C₁₀H₁₆O₄
Melting Pt: 183…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00001375
CAS Number: 124-83-4
EINECS: 204-715-0
Merck Index: 14,1733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR