9,10-Dimethylanthracene ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B22458.03 B22458.06
B22458.03EA 75.8 GBP
B22458.03 B22458.06
9,10-Dimethylanthracene ≥99%
9,10-Dimethylanthracene

Purity: 99% CAS No.: 781-43-1 Molecular Formula: C16H14 Formula Weight: 206.29 Melting Point: 182-184° Beilstein Reference: 1909028

Formula: C₁₆H₁₄
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00001262
CAS Number: 781-43-1
EINECS: 212-308-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR