Azo rubine

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B22328.09 B22328.18 B22328.30
B22328.09EA 26.2 GBP
B22328.09 B22328.18 B22328.30
Azo rubine
Azo rubine

CAS No.: 3567-69-9 Molecular Formula: C20H12N2Na2O7S2 Formula Weight: 502.42 MDL No.: MFCD00003978

Formula: C₂₀H₁₄N₂O₇S₂
Storage Temperature: Ambient
MDL Number: MFCD00003978
CAS Number: 3567-69-9
EINECS: 222-657-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR