Toluene-α-sulphonyl fluoride 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

B22146.06 B22146.03 B22146.14
B22146.06EA 43.2 GBP
B22146.06 B22146.03 B22146.14
Toluene-α-sulphonyl fluoride 99
Toluene-α-sulphonyl fluoride

Purity: 99% CAS No.: 329-98-6 Molecular Formula: C7H7FO2S Formula Weight: 174.19 Melting Point: 92-94° Beilstein Reference: 2088311 UN No.: 2923 MDL No.: MFCD00007424

Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Boiling Pt: 80…85 °C (0.3 mmHg)
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR