Calix[8]arene ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B21666.03 B21666.06 B21666.14
B21666.03EA 48.4 GBP
B21666.03 B21666.06 B21666.14
Calix[8]arene ≥97%
Calix[8]arene

Purity: 97% CAS No.: 82452-93-5 Molecular Formula: C56H48O8 Formula Weight: 848.99 Melting Point: 450° MDL No.: MFCD00143914

Formula: C₅₆H₄₈O₈
MW: 848.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143914
CAS Number: 82452-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR