L(+)-Asparagine, anhydrous 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: L-Asparagine, (S)-(+)-Asparagine, H-Asn-OH

B21473.0E B21473.22 B21473.36
B21473.0EEA 651 GBP
B21473.0E B21473.22 B21473.36
L(+)-Asparagine, anhydrous 99%
L(+)-Asparagine

Purity: 99% CAS No.: 70-47-3 Molecular Formula: C4H8N2O3 Formula Weight: 132.12 Melting Point: 234-236° dec. Beilstein Reference: 1723527 Merck Reference: 14,837 MDL No.: MFCD00064401

Formula: H₂NCOCH₂CH(NH₂)CO₂H
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 234…236 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064401
CAS Number: 70-47-3
EINECS: 200-735-9
Merck Index: 14,837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=3d09743d0834d64e4719e8295f634c39195c571f&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR