4-tert-Butylcalix[8]arene 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B21443.03 B21443.06 B21443.14
B21443.03EA 17.3 GBP
B21443.03 B21443.06 B21443.14
4-tert-Butylcalix[8]arene 96%
4-tert-Butylcalix[8]arene

Purity: 96% CAS No.: 68971-82-4 Molecular Formula: C88H112O8 Formula Weight: 1297.87 Melting Point: 411-412° Beilstein Reference: 2612052

Formula: C₈₈H₁₁₂O₈
MW: 1297.85 g/mol
Melting Pt: 411…412 °C
Storage Temperature: Ambient
MDL Number: MFCD00075392
CAS Number: 68971-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR