Tetracycline hydrochloride 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

B21408.14 B21408.22
B21408.14EA 34.9 GBP
B21408.14 B21408.22
Tetracycline hydrochloride 96%
Tetracycline hydrochloride

Purity: 96% CAS No.: 64-75-5 Molecular Formula: C22H24N2O8.HCl Formula Weight: 480.91 Melting Point: 220-223° Merck Reference: 14,9196 Optical Rotation: -247° (c=1, 0.1M HCl) MDL No.: MFCD00078142


Can induce apoptosis in osteoclasts

Formula: C₂₂H₂₅ClN₂O₈
Melting Pt: 220…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8
Merck Index: 14,9196

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR